siRNA Details
| Gene Symbol: KRT34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021013.3 | NP_066293.2 | 7 | 1270 | ||
| XM_011524793.1 | XP_011523095.1 | 8 | 1349 | |||
| GenBank mRNA | BC033252.1 | - | 5 | 607 | ||
| BC041070.1 | - | 7 | 1289 | 5481838 | ||

KRT34
Genomic Map