siRNA Details
| Gene Symbol: MAGEA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011543.2 | NP_001011543.2 | 5 | 1392 | ||
| NM_001251828.1 | NP_001238757.1 | 5 | 1320 | |||
| NM_021048.4 | NP_066386.2 | 4 | 1301 | |||
| GenBank mRNA | AK314161.1 | - | 4 | 1301 | ||
| BC004105.2 | - | 4 | 1276 | 3926146 | ||

MAGEA10
Genomic Map