siRNA Details
| Gene Symbol: NAP1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021963.3 | NP_068798.1 | 1 | 1323 | ||
| GenBank mRNA | AB027013.1 | - | 1 | 1297 | ||
| AF136178.1 | - | 1 | 1271 | |||
| AK291297.1 | - | 1 | 1281 | |||
| AK303676.1 | - | 2 | 916 | |||
| AK316569.1 | - | 1 | 1323 | |||
| BC026325.1 | - | 1 | 1245 | 4798964 | ||
| CR457095.1 | - | 1 | 941 | |||

NAP1L2
Genomic Map