siRNA Details
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058004.3 | NP_477352.3 | 42, 43 | 5080 | ||
| XM_005261634.1 | XP_005261691.1 | 41, 42 | 4987 | |||
| XM_017028829.1 | XP_016884318.1 | 41, 42 | 5014 | |||
| XM_017028830.1 | XP_016884319.1 | 28, 29 | 3236 | |||
| GenBank mRNA | AB210002.1 | - | 42, 43 | 5050 | ||
| AF012872.1 | - | 41, 42 | 4819 | |||
| AK302424.1 | - | 42, 43 | 5008 | |||
| AK314781.1 | - | 14, 15 | 1734 | |||
| BC018120.1 | - | 12, 13 | 1397 | |||
| BC053654.1 | - | 11, 12 | 1373 | |||
| L36151.1 | - | 10, 11 | 1366 | |||

PI4KA
Genomic Map