siRNA Details
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 6 | 2000 | ||
| NM_153818.1 | NP_722540.1 | 6 | 2060 | |||
| GenBank mRNA | BC000543.2 | - | 6 | 2019 | 3163782 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PEX10| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 6 | 2000 | ||
| NM_153818.1 | NP_722540.1 | 6 | 2060 | |||
| GenBank mRNA | BC000543.2 | - | 6 | 2019 | 3163782 | |