siRNA Details
| Gene Symbol: NHS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136024.3 | NP_001129496.1 | 3 | 585 | ||
| NM_001291867.1 | NP_001278796.1 | 3 | 1180 | |||
| NM_001291868.1 | NP_001278797.1 | 3 | 585 | |||
| NM_198270.3 | NP_938011.1 | 3 | 1180 | |||
| GenBank mRNA | AK302437.1 | - | 3 | 1366 | ||
| AY436752.1 | - | 3 | 1180 | |||
| AY456992.1 | - | 3 | 585 | |||
| AY456993.1 | - | 2 | 1240 | |||
| BC136415.1 | - | 3 | 906 | 9020403 | ||
| BC171763.1 | - | 3 | 906 | 9054416 | ||
| CR936788.1 | - | 3 | 354 | |||
| GQ988776.1 | - | 3 | 842 | |||

NHS
Genomic Map