siRNA Details
| Gene Symbol: FADS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281501.1 | NP_001268430.1 | 3 | 388 | ||
| NM_001281502.1 | NP_001268431.1 | 3 | 457 | |||
| NM_004265.3 | NP_004256.1 | 3 | 729 | |||
| GenBank mRNA | AF084559.1 | - | 3 | 521 | ||
| AF108658.1 | - | 3 | 314 | |||
| AF126799.1 | - | 3 | 384 | |||
| AK074939.1 | - | 3 | 461 | |||
| AK074991.1 | - | 3 | 197 | |||
| AK290291.1 | - | 3 | 335 | |||
| AK299762.1 | - | 3 | 457 | |||
| BC009011.1 | - | 3 | 463 | 4181037 | ||
| HY090295.1 | - | 3 | 389 | |||

FADS2
Genomic Map