siRNA Details
| Gene Symbol: GRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278064.1 | NP_001264993.1 | 2 | 1082 | ||
| NM_001278065.1 | NP_001264994.1 | 3 | 1129 | |||
| NM_001278066.1 | NP_001264995.1 | 2 | 1082 | |||
| NM_001278067.1 | NP_001264996.1 | 2 | 1082 | |||
| XM_011535782.1 | XP_011534084.1 | 3 | 1232 | |||
| XM_017010783.1 | XP_016866272.1 | 3 | 1241 | |||
| XM_017010784.1 | XP_016866273.1 | 3 | 1277 | |||
| XM_017010785.1 | XP_016866274.1 | 3 | 1237 | |||
| XM_017010786.1 | XP_016866275.1 | 3 | 1237 | |||
| XM_017010787.1 | XP_016866276.1 | 3 | 1273 | |||
| GenBank mRNA | AB208837.1 | - | 2 | 749 | ||
| AK308550.1 | - | 3 | 1232 | |||
| BC136280.1 | - | 2 | 794 | 9020267 | ||
| BC143777.1 | - | 2 | 794 | |||
| BC143779.1 | - | 2 | 792 | |||
| L76627.1 | - | 2 | 762 | |||
| L76631.1 | - | 3 | 1132 | |||
| U31215.1 | - | 2 | 997 | |||
| U31216.1 | - | 2 | 762 | |||
More Information
Set Membership: |
|

GRM1
Genomic Map