siRNA Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004278.3 | NP_004269.1 | 2 | 293 | ||
| XM_011524080.2 | XP_011522382.1 | 2 | 279 | |||
| XM_017025349.1 | XP_016880838.1 | 2 | 279 | |||
| XM_017025350.1 | XP_016880839.1 | 2 | 279 | |||
| XM_017025351.1 | XP_016880840.1 | 2 | 279 | |||
| XM_017025352.1 | XP_016880841.1 | 2 | 279 | |||
| XM_017025353.1 | XP_016880842.1 | 2 | 279 | |||
| XM_017025354.1 | XP_016880843.1 | 2 | 279 | |||
| XM_017025355.1 | XP_016880844.1 | 2 | 279 | |||
| XM_017025356.1 | XP_016880845.1 | 2 | 279 | |||
| GenBank mRNA | AB017165.1 | - | 2 | 266 | ||
| AK292932.1 | - | 2 | 278 | |||
| AK302523.1 | - | 2 | 278 | |||
| BC068197.1 | - | 2 | 272 | 6711166 | ||

PIGL
Genomic Map