siRNA Details
| Gene Symbol: HTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000870.6 | NP_000861.1 | 6 | 1354 | ||
| NM_001040169.2 | NP_001035259.1 | 5 | 888 | |||
| NM_001040172.2 | NP_001035262.2 | 5 | 888 | |||
| NM_001040173.2 | NP_001035263.1 | 5 | 888 | |||
| NM_001286410.1 | NP_001273339.1 | 5 | 888 | |||
| NM_199453.3 | NP_955525.1 | 5 | 888 | |||
| NR_104445.1 | 6 | 1354 | ||||
| GenBank mRNA | AJ011371.1 | - | 5 | 843 | ||
| AJ131724.1 | - | 6 | 981 | |||
| AJ131725.1 | - | 5 | 847 | |||
| AJ131726.1 | - | 5 | 847 | |||
| AJ278979.1 | - | 5 | 847 | |||
| AJ278980.1 | - | 5 | 847 | |||
| AJ278981.1 | - | 5 | 847 | |||
| AJ278982.1 | - | 5 | 847 | |||
| AJ519673.1 | - | 5 | 860 | |||
| AJ633645.1 | - | 5 | 841 | |||
| AK308471.1 | - | 6 | 1354 | |||
| AM712912.1 | - | 5 | 841 | |||
| BC074755.2 | - | 5 | 882 | 30915214 | ||
| BC095497.1 | - | 5 | 847 | 6971934 | ||
| Y08756.1 | - | 5 | 859 | |||
| Y09586.1 | - | 5 | 843 | |||
| Y10437.1 | - | 5 | 841 | |||
| Y12505.1 | - | 5 | 859 | |||
| Y12506.1 | - | 5 | 859 | |||
| Y12507.1 | - | 5 | 859 | |||
| Y13584.1 | - | 5 | 841 | |||
More Information
Set Membership: |
|

HTR4
Genomic Map