siRNA Details
| Gene Symbol: FLNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127487.1 | NP_001120959.1 | 30 | 5308 | ||
| NM_001458.4 | NP_001449.3 | 30 | 5308 | |||
| GenBank mRNA | AB208865.1 | - | 7 | 950 | ||
| AB371585.1 | - | 30 | 5328 | |||
| AF089841.1 | - | 30 | 5148 | |||
| AF146692.1 | - | 30 | 5311 | |||
| AJ012737.1 | - | 30 | 5117 | |||
| X70084.1 | - | 2 | 190 | |||

FLNC
Genomic Map