siRNA Details
| Gene Symbol: GUCY2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001522.2 | NP_001513.2 | 2 | 536 | ||
| GenBank mRNA | BC144241.1 | - | 1 | 331 | ||
| L37378.1 | - | 2 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY2F| Gene Symbol: GUCY2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001522.2 | NP_001513.2 | 2 | 536 | ||
| GenBank mRNA | BC144241.1 | - | 1 | 331 | ||
| L37378.1 | - | 2 | 559 | |||