siRNA Details
| Gene Symbol: EIF3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001568.2 | NP_001559.1 | 3 | 260 | ||
| XM_017013398.1 | XP_016868887.1 | 3 | 332 | |||
| GenBank mRNA | AK124178.1 | - | 3 | 864 | ||
| AK310902.1 | - | 3 | 332 | |||
| AK313186.1 | - | 3 | 249 | |||
| BC000734.2 | - | 3 | 236 | 2959855 | ||
| BC005944.1 | - | 3 | 249 | |||
| BC008419.1 | - | 3 | 254 | 4080571 | ||
| BC016706.1 | - | 3 | 251 | 4096434 | ||
| BC017887.1 | - | 3 | 256 | 4685684 | ||
| BC021679.1 | - | 3 | 251 | 4132737 | ||
| BG498300.1 | - | 3 | 261 | |||
| CR457029.1 | - | 3 | 232 | |||
| CR542275.1 | - | 3 | 232 | |||
| KC333871.1 | - | 3 | 260 | |||
| U54562.1 | - | 3 | 238 | |||
| U62962.1 | - | 3 | 254 | |||
| U85947.1 | - | 3 | 240 | |||
| U94175.1 | - | 3 | 252 | |||

EIF3E
Genomic Map