siRNA Details
| Gene Symbol: IRAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025242.1 | NP_001020413.1 | 5 | 782 | ||
| NM_001025243.1 | NP_001020414.1 | 5 | 782 | |||
| NM_001569.3 | NP_001560.2 | 5 | 782 | |||
| XM_005274668.3 | XP_005274725.1 | 4 | 804 | |||
| XM_011531158.2 | XP_011529460.1 | 5 | 797 | |||
| GenBank mRNA | AF346607.1 | - | 5 | 703 | ||
| BC014963.1 | - | 5 | 767 | 4906853 | ||
| BC054000.1 | - | 4 | 859 | 6164719 | ||
| DQ054788.1 | - | 5 | 703 | |||
| L76191.1 | - | 5 | 782 | |||

IRAK1
Genomic Map