siRNA Details
| Gene Symbol: FPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002030.4 | NP_002021.3 | 2 | 1427 | ||
| XM_011526687.2 | XP_011524989.1 | 2 | 1318 | |||
| GenBank mRNA | AK123030.1 | - | 3 | 1370 | ||
| BC059388.1 | - | 2 | 1410 | 30332008 | ||
More Information
Set Membership: |
|

FPR3
Genomic Map