siRNA Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002077.3 | NP_002068.1 | 6 | 693 | ||
| XM_005251929.3 | XP_005251986.1 | 6 | 625 | |||
| XM_006717062.3 | XP_006717125.1 | 6 | 689 | |||
| XM_006717063.3 | XP_006717126.1 | 6 | 660 | |||
| GenBank mRNA | AB208858.1 | - | 1 | 50 | ||
| AK302445.1 | - | 6 | 502 | |||
| AK307789.1 | - | 3 | 360 | |||
| BC032853.1 | - | 6 | 663 | 4828793 | ||
| BG699932.1 | - | 6 | 668 | |||
| U51587.1 | - | 6 | 685 | |||

GOLGA1
Genomic Map