siRNA Details
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002190.2 | NP_002181.1 | 3 | 1186 | ||
GenBank mRNA | U32659.1 | - | 3 | 1194 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002190.2 | NP_002181.1 | 3 | 1186 | ||
GenBank mRNA | U32659.1 | - | 3 | 1194 |