siRNA Details
| Gene Symbol: INHBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002193.2 | NP_002184.2 | 2 | 2513 | ||
| GenBank mRNA | BC030029.2 | - | 2 | 2513 | 5258851 | |
| BC127815.1 | - | 1 | 250 | |||
| CK430845.1 | - | 1 | 480 | |||
| M31682.1 | - | 2 | 2407 | |||

INHBB
Genomic Map