siRNA Details
| Gene Symbol: ITGAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002208.4 | NP_002199.3 | 8, 9 | 957 | ||
| XM_011523823.2 | XP_011522125.1 | 8, 9 | 1395 | |||
| XM_011523825.2 | XP_011522127.1 | 8, 9 | 1393 | |||
| XM_011523827.2 | XP_011522129.1 | 8, 9 | 1387 | |||
| XM_011523828.2 | XP_011522130.1 | 8, 9 | 1391 | |||
| XM_017024586.1 | XP_016880075.1 | 9, 10 | 2780 | |||
| XM_017024587.1 | XP_016880076.1 | 9, 10 | 2780 | |||
| GenBank mRNA | BC113436.1 | - | 8, 9 | 947 | 8322488 | |
| BC117207.1 | - | 8, 9 | 947 | 40125758 | ||
| L25851.2 | - | 8, 9 | 983 | |||

ITGAE
Genomic Map