siRNA Details
| Gene Symbol: ITGAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002208.4 | NP_002199.3 | 10 | 1211 | ||
| XM_011523823.2 | XP_011522125.1 | 10 | 1649 | |||
| XM_011523825.2 | XP_011522127.1 | 10 | 1647 | |||
| XM_011523827.2 | XP_011522129.1 | 10 | 1641 | |||
| XM_011523828.2 | XP_011522130.1 | 10 | 1645 | |||
| XM_017024586.1 | XP_016880075.1 | 11 | 3034 | |||
| XM_017024587.1 | XP_016880076.1 | 11 | 3034 | |||
| GenBank mRNA | BC113436.1 | - | 10 | 1201 | 8322488 | |
| BC117207.1 | - | 10 | 1201 | 40125758 | ||
| L25851.2 | - | 10 | 1237 | |||

ITGAE
Genomic Map