siRNA Details
| Gene Symbol: ITIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002216.2 | NP_002207.2 | 6 | 698 | ||
| GenBank mRNA | AK225277.1 | - | 8 | 561 | ||
| AK225329.1 | - | 6 | 603 | |||
| AK291379.1 | - | 6 | 646 | |||
| BC132685.1 | - | 6 | 585 | 40146707 | ||
| X07173.1 | - | 6 | 599 | |||

ITIH2
Genomic Map