siRNA Details
| Gene Symbol: ITPKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002220.2 | NP_002211.1 | 1, 2 | 558 | ||
| GenBank mRNA | BC026331.1 | - | 1, 2 | 542 | 4792595 | |
| X54938.1 | - | 1, 2 | 499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPKA| Gene Symbol: ITPKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002220.2 | NP_002211.1 | 1, 2 | 558 | ||
| GenBank mRNA | BC026331.1 | - | 1, 2 | 542 | 4792595 | |
| X54938.1 | - | 1, 2 | 499 | |||