siRNA Details
| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002221.3 | NP_002212.3 | 2 | 1240 | ||
| XM_005273120.2 | XP_005273177.1 | 2 | 1392 | |||
| XM_017001211.1 | XP_016856700.1 | 2 | 1392 | |||
| GenBank mRNA | BC015009.1 | - | 2 | 1240 | 4862726 | |
| Y18024.2 | - | 1 | 900 | |||

ITPKB
Genomic Map