siRNA Details
| Gene Symbol: UBE2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004223.4 | NP_004214.1 | 4 | 991 | ||
| NM_198183.2 | NP_937826.1 | 4 | 1350 | |||
| GenBank mRNA | AF061736.1 | - | 4 | 942 | ||
| AK093462.1 | - | 4 | 1304 | |||
| AK129621.1 | - | 4 | 862 | |||
| AK226165.1 | - | 4 | 1045 | |||
| BC032491.1 | - | 4 | 991 | 5246109 | ||

UBE2L6
Genomic Map