siRNA Details
| Gene Symbol: GPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005282.2 | NP_005273.1 | 2 | 3004 | ||
| XM_017026607.1 | XP_016882096.1 | 2 | 3592 | |||
| XM_017026608.1 | XP_016882097.1 | 2 | 2913 | |||
| GenBank mRNA | BC038450.1 | - | 2 | 2991 | ||

GPR4
Genomic Map