siRNA Details
| Gene Symbol: GPR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161415.1 | NP_001154887.1 | 4 | 1747 | ||
| NM_001161416.1 | NP_001154888.1 | 3 | 1682 | |||
| NM_001161417.1 | NP_001154889.1 | 2 | 1453 | |||
| NM_005291.2 | NP_005282.1 | 3 | 1518 | |||
| XM_017003833.1 | XP_016859322.1 | 2 | 1874 | |||
| GenBank mRNA | AK126849.1 | - | 1 | 2708 | ||
| AK292725.1 | - | 2 | 1323 | |||
| BC031653.1 | - | 2 | 1132 | 5174146 | ||
| BC039595.1 | - | 2 | 1137 | |||
| Y12546.1 | - | 3 | 1210 | |||
| Z94154.1 | - | 2 | 1146 | |||
| Z94155.1 | - | 1 | 930 | |||

GPR17
Genomic Map