siRNA Details
| Gene Symbol: GPR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195381.1 | NP_001182310.1 | 6 | 1970 | ||
| NM_001195382.1 | NP_001182311.1 | 6 | 1856 | |||
| NM_005301.3 | NP_005292.2 | 2 | 1333 | |||
| GenBank mRNA | AF089087.1 | - | 1 | 1580 | ||
| AK131540.1 | - | 1 | 3516 | |||
| AK172786.1 | - | 6 | 1947 | |||
| AK290256.1 | - | 6 | 1819 | |||
| BC117453.1 | - | 1 | 1377 | 40126004 | ||
| BC117455.1 | - | 1 | 1375 | 40126006 | ||

GPR35
Genomic Map