siRNA Details
| Gene Symbol: FFAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005306.2 | NP_005297.1 | 1 | 514 | ||
| XM_017026709.1 | XP_016882198.1 | 3 | 1053 | |||
| XM_017026710.1 | XP_016882199.1 | 2 | 930 | |||
| XM_017026711.1 | XP_016882200.1 | 2 | 688 | |||
| GenBank mRNA | AB378083.1 | - | 1 | 514 | ||
| AX810701.1 | - | 2 | 594 | |||
| BC095535.1 | - | 1 | 511 | |||
| BC096198.3 | - | 1 | 514 | 40004248 | ||
| BC096199.3 | - | 1 | 514 | 40004249 | ||
| BC096200.1 | - | 1 | 514 | |||
| BC096201.1 | - | 1 | 514 | 40004253 | ||

FFAR2
Genomic Map