siRNA Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301228.1 | NP_001288157.1 | 5 | 1245 | ||
| NM_001301229.1 | NP_001288158.1 | 5 | 1136 | |||
| NM_001301231.1 | NP_001288160.1 | 5 | 1175 | |||
| NM_005342.3 | NP_005333.2 | 5 | 1140 | |||
| GenBank mRNA | BX537505.1 | - | 3 | 2536 | ||
| Y10043.1 | - | 5 | 1119 | |||

HMGB3
Genomic Map