siRNA Details
| Gene Symbol: LRRC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 2332 | ||
| NM_005512.2 | NP_005503.1 | 3 | 2437 | |||
| XM_005273903.4 | XP_005273960.2 | 3 | 2419 | |||
| XM_011544935.2 | XP_011543237.1 | 3 | 2728 | |||
| GenBank mRNA | BC052210.1 | - | 3 | 2329 | 4825609 | |
| BC070079.1 | - | 3 | 2332 | 30344529 | ||
| DA591993.1 | - | 1 | 229 | |||
| Z24680.1 | - | 3 | 2287 | |||

LRRC32
Genomic Map