siRNA Details
| Gene Symbol: HSPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291860.1 | NP_001278789.1 | 2 | 219 | ||
| NM_005529.6 | NP_005520.4 | 2 | 219 | |||
| XM_011541318.2 | XP_011539620.1 | 2 | 154 | |||
| XM_017001120.1 | XP_016856609.1 | 2 | 295 | |||
| XM_017001121.1 | XP_016856610.1 | 2 | 295 | |||
| XM_017001122.1 | XP_016856611.1 | 2 | 295 | |||
| GenBank mRNA | AF479675.1 | - | 1 | 79 | ||
| M85289.1 | - | 2 | 219 | |||
| X62515.1 | - | 2 | 179 | |||

HSPG2
Genomic Map