siRNA Details
| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265587.1 | NP_001252516.1 | 3 | 642 | ||
| NM_005543.3 | NP_005534.2 | 2 | 547 | |||
| GenBank mRNA | AK302780.1 | - | 2 | 489 | ||
| AY082014.1 | - | 3 | 642 | |||
| BC032810.1 | - | 2 | 477 | |||
| BC040435.1 | - | 2 | 477 | |||
| BC053345.1 | - | 2 | 477 | |||
| BC061886.1 | - | 2 | 500 | |||
| BC071706.1 | - | 2 | 480 | 5167223 | ||
| BC106721.1 | - | 2 | 479 | 40014433 | ||
| BC106722.1 | - | 2 | 484 | 40014435 | ||
| BC107045.2 | - | 1 | 173 | |||
| BX334591.2 | - | 2 | 491 | |||
| S72482.1 | - | 2 | 486 | |||

INSL3
Genomic Map