siRNA Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145349.1 | NP_001138821.1 | 3, 4 | 999 | ||
| NM_005897.2 | NP_005888.1 | 3, 4 | 999 | |||
| XM_005270839.4 | XP_005270896.1 | 3, 4 | 999 | |||
| XM_006710623.3 | XP_006710686.1 | 3, 4 | 999 | |||
| XM_011541394.2 | XP_011539696.1 | 3, 4 | 999 | |||
| XM_017001210.1 | XP_016856699.1 | 3, 4 | 999 | |||
| GenBank mRNA | AF156857.1 | - | 2, 3 | 782 | ||
| AK023880.1 | - | 3, 4 | 835 | |||
| BC032544.1 | - | 3, 4 | 840 | 5535760 | ||

IPP
Genomic Map