siRNA Details
| Gene Symbol: SFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006142.3 | NP_006133.1 | 1 | 502 | ||
| GenBank mRNA | AF029082.1 | - | 1 | 502 | ||
| BC000329.2 | - | 1 | 482 | 2822397 | ||
| BC000995.2 | - | 1 | 479 | 3445915 | ||
| BC001550.1 | - | 2 | 388 | 3457665 | ||
| BC002995.1 | - | 1 | 482 | 2822397 | ||
| BC023552.2 | - | 1 | 506 | 3534328 | ||
| CR541905.1 | - | 1 | 431 | |||
| CR541926.1 | - | 1 | 431 | |||
| M93010.1 | - | 1 | 441 | |||

SFN
Genomic Map