siRNA Details
| Gene Symbol: CENPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318521.1 | NP_001305450.1 | 20 | 2085 | ||
| NM_006733.3 | NP_006724.2 | 19 | 2179 | |||
| XM_005262111.2 | XP_005262168.1 | 20 | 2049 | |||
| XM_011530894.2 | XP_011529196.1 | 20 | 2078 | |||
| XM_011530895.2 | XP_011529197.1 | 21 | 2365 | |||
| XM_011530897.2 | XP_011529199.1 | 21 | 2240 | |||
| XM_011530899.2 | XP_011529201.1 | 19 | 2046 | |||
| XM_017029383.1 | XP_016884872.1 | 20 | 2078 | |||
| GenBank mRNA | AK302986.1 | - | 20 | 2078 | ||
| BG114761.1 | - | 330 | ||||
| X97249.1 | - | 19 | 2178 | |||

CENPI
Genomic Map