siRNA Details
| Gene Symbol: COX5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004255.3 | NP_004246.2 | 4 | 532 | ||
| GenBank mRNA | AK026615.1 | - | 4 | 422 | ||
| AK026623.1 | - | 4 | 426 | |||
| BC024240.2 | - | 4 | 506 | 4815806 | ||
| CR407649.1 | - | 4 | 392 | |||
| DQ987236.1 | - | 4 | 392 | |||
| DQ987237.1 | - | 4 | 392 | |||
| M22760.1 | - | 4 | 411 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007747, BC034302, X15963, AK018723, BC003831.

COX5A
Genomic Map