siRNA Details
| Gene Symbol: HIST1H2AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021065.3 | NP_066409.1 | 1 | 454 | ||
| GenBank mRNA | BC093807.1 | - | 1 | 443 | 7939652 | |
| BC093809.1 | - | 1 | 446 | 7939654 | ||
| BC128035.1 | - | 1 | 404 | 40021702 | ||

HIST1H2AD
Genomic Map