siRNA Details
| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004256.3 | NP_004247.2 | 10 | 2202 | ||
| XM_017007519.1 | XP_016863008.1 | 9 | 1948 | |||
| GenBank mRNA | AB010438.1 | - | 10 | 2169 | ||
| BC035973.1 | - | 10 | 2175 | 4609915 | ||

SLC22A13
Genomic Map