siRNA Details
| Gene Symbol: HNRNPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004501.3 | NP_004492.2 | 4 | 1142 | ||
| NM_031844.2 | NP_114032.2 | 4 | 1199 | |||
| XM_017001115.1 | XP_016856604.1 | 4 | 1394 | |||
| XM_017001116.1 | XP_016856605.1 | 4 | 1199 | |||
| XM_017001117.1 | XP_016856606.1 | 4 | 1188 | |||
| GenBank mRNA | AF068846.1 | - | 4 | 1195 | ||
| AF461014.1 | - | 1 | 720 | |||
| AK126868.1 | - | 5 | 940 | |||
| AK130669.1 | - | 3 | 248 | |||
| AK297119.1 | - | 5 | 1033 | |||
| AK307661.1 | - | 4 | 666 | |||
| BC003367.1 | - | 4 | 1073 | 2900876 | ||
| BC003621.2 | - | 4 | 1075 | 2966453 | ||
| BC007950.2 | - | 4 | 623 | |||
| BC015782.1 | - | 4 | 676 | 4857112 | ||
| BC024767.1 | - | 4 | 1075 | 4993703 | ||
| BC034925.1 | - | 1 | 66 | |||
| X65488.1 | - | 4 | 965 | |||

HNRNPU
Genomic Map