siRNA Details
| Gene Symbol: JCHAIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144646.3 | NP_653247.1 | 4 | 514 | ||
| XM_006714211.2 | XP_006714274.1 | 5 | 479 | |||
| XM_006714212.2 | XP_006714275.1 | 5 | 416 | |||
| XM_011531925.1 | XP_011530227.1 | 5 | 416 | |||
| XM_011531926.1 | XP_011530228.1 | 5 | 416 | |||
| GenBank mRNA | AK312014.1 | - | 4 | 404 | ||
| BC038982.1 | - | 4 | 430 | 5755337 | ||
| BP261870.1 | - | 4 | 543 | |||
| BX648421.1 | - | 1 | 1974 | |||
| DQ884395.1 | - | 5 | 421 | |||

JCHAIN
Genomic Map