siRNA Details
| Gene Symbol: STT3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278503.1 | NP_001265432.1 | 11 | 1322 | ||
| NM_001278504.1 | NP_001265433.1 | 9 | 1111 | |||
| NM_152713.4 | NP_689926.1 | 10 | 1234 | |||
| XM_011542807.2 | XP_011541109.1 | 10 | 1261 | |||
| GenBank mRNA | AK290040.1 | - | 10 | 1184 | ||
| AK290657.1 | - | 10 | 1182 | |||
| AK295892.1 | - | 9 | 1039 | |||
| AK308902.1 | - | 10 | 1177 | |||
| BC020965.2 | - | 10 | 1167 | 3891543 | ||
| BC048348.1 | - | 9 | 1120 | 5561615 | ||
| BT007100.1 | - | 9 | 1075 | |||
| BX649102.1 | - | 11 | 1260 | |||
| L38961.1 | - | 10 | 1181 | |||
| L47337.1 | - | 10 | 1191 | |||

STT3A
Genomic Map