siRNA Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000167.5 | NP_000158.1 | 19 | 1993 | ||
| NM_001128127.2 | NP_001121599.1 | 20 | 2080 | |||
| NM_001205019.1 | NP_001191948.1 | 21 | 2098 | |||
| NM_203391.3 | NP_976325.1 | 20 | 2011 | |||
| XM_005274488.4 | XP_005274545.1 | 18 | 1850 | |||
| XM_006724483.2 | XP_006724546.1 | 21 | 2117 | |||
| XM_006724484.2 | XP_006724547.1 | 20 | 2029 | |||
| XM_006724485.2 | XP_006724548.1 | 20 | 1943 | |||
| XM_006724486.3 | XP_006724549.1 | 19 | 1928 | |||
| XM_011545491.2 | XP_011543793.1 | 22 | 2136 | |||
| XM_011545492.1 | XP_011543794.1 | 21 | 2048 | |||
| XM_011545493.2 | XP_011543795.1 | 20 | 1818 | |||
| XM_011545494.2 | XP_011543796.1 | 20 | 1763 | |||
| XM_017029409.1 | XP_016884898.1 | 22 | 2133 | |||
| XM_017029410.1 | XP_016884899.1 | 18 | 1825 | |||
| XM_017029411.1 | XP_016884900.1 | 18 | 1840 | |||
| XM_017029412.1 | XP_016884901.1 | 17 | 1761 | |||
| GenBank mRNA | AK294304.1 | - | 21 | 1817 | ||
| AK294938.1 | - | 22 | 2133 | |||
| BC042421.1 | - | 20 | 2011 | 4820945 | ||
| BC071595.1 | - | 20 | 2011 | 5276056 | ||
| BX647604.1 | - | 20 | 1832 | |||
| L13943.1 | - | 19 | 1879 | |||
| X69886.1 | - | 15 | 1431 | |||

GK
Genomic Map