siRNA Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000167.5 | NP_000158.1 | 12 | 1124 | ||
| NM_001128127.2 | NP_001121599.1 | 12 | 1124 | |||
| NM_001205019.1 | NP_001191948.1 | 13 | 1142 | |||
| NM_203391.3 | NP_976325.1 | 13 | 1142 | |||
| XM_005274488.4 | XP_005274545.1 | 10 | 894 | |||
| XM_006724483.2 | XP_006724546.1 | 13 | 1161 | |||
| XM_006724484.2 | XP_006724547.1 | 13 | 1160 | |||
| XM_006724485.2 | XP_006724548.1 | 12 | 987 | |||
| XM_006724486.3 | XP_006724549.1 | 11 | 972 | |||
| XM_011545491.2 | XP_011543793.1 | 14 | 1180 | |||
| XM_011545492.1 | XP_011543794.1 | 14 | 1179 | |||
| XM_011545493.2 | XP_011543795.1 | 12 | 862 | |||
| XM_011545494.2 | XP_011543796.1 | 12 | 807 | |||
| XM_017029409.1 | XP_016884898.1 | 14 | 1177 | |||
| XM_017029410.1 | XP_016884899.1 | 11 | 956 | |||
| XM_017029411.1 | XP_016884900.1 | 11 | 971 | |||
| XM_017029412.1 | XP_016884901.1 | 10 | 892 | |||
| GenBank mRNA | AK294304.1 | - | 13 | 948 | ||
| AK294938.1 | - | 14 | 1177 | |||
| AK308238.1 | - | 10 | 803 | |||
| AK313215.1 | - | 12 | 1060 | |||
| BC037549.1 | - | 12 | 981 | 5213716 | ||
| BC042421.1 | - | 13 | 1143 | 4820945 | ||
| BC071595.1 | - | 13 | 1143 | 5276056 | ||
| BX647604.1 | - | 13 | 963 | |||
| L13943.1 | - | 12 | 1011 | |||
| X68285.1 | - | 11 | 735 | |||
| X69886.1 | - | 8 | 560 | |||

GK
Genomic Map