siRNA Details
| Gene Symbol: COL1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000088.3 | NP_000079.2 | 11, 12 | 939 | ||
| XM_005257058.3 | XP_005257115.2 | 11, 12 | 948 | |||
| XM_005257059.4 | XP_005257116.2 | 11, 12 | 948 | |||
| XM_011524341.1 | XP_011522643.1 | 11, 12 | 948 | |||
| GenBank mRNA | AB209597.1 | - | 11, 12 | 914 | ||
| BC036531.2 | - | 11, 12 | 929 | 5264710 | ||
| JQ236861.1 | - | 11, 12 | 813 | |||
| M36546.1 | - | 11, 12 | 896 | |||
| X07884.1 | - | 11, 12 | 896 | |||
| Z74615.1 | - | 11, 12 | 932 | |||

COL1A1
Genomic Map