siRNA Details
| Gene Symbol: CCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123041.2 | NP_001116513.2 | 2 | 1079 | ||
| NM_001123396.1 | NP_001116868.1 | 2 | 1079 | |||
| XM_011534069.1 | XP_011532371.1 | 3 | 792 | |||
| GenBank mRNA | AK292685.1 | - | 2 | 1089 | ||
| AK292920.1 | - | 2 | 697 | |||
| BC074751.2 | - | 1 | 633 | 30915210 | ||
| BC095540.1 | - | 1 | 598 | 7001971 | ||
| BC126452.1 | - | 1 | 628 | 8992168 | ||
| BC136396.1 | - | 1 | 628 | 9020383 | ||
| D29984.1 | - | 1 | 594 | |||
| U03882.1 | - | 1 | 633 | |||
| U03905.1 | - | 2 | 674 | |||

CCR2
Genomic Map