siRNA Details
| Gene Symbol: CD3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000733.3 | NP_000724.1 | 9 | 1104 | ||
| GenBank mRNA | AK292612.1 | - | 9 | 953 | ||
| BC049847.1 | - | 8 | 916 | 5214563 | ||
| X03884.1 | - | 8 | 902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD3E| Gene Symbol: CD3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000733.3 | NP_000724.1 | 9 | 1104 | ||
| GenBank mRNA | AK292612.1 | - | 9 | 953 | ||
| BC049847.1 | - | 8 | 916 | 5214563 | ||
| X03884.1 | - | 8 | 902 | |||