siRNA Details
| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244.3 | NP_001235.1 | 4 | 1208 | ||
| GenBank mRNA | L09753.1 | - | 4 | 1030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFSF8| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244.3 | NP_001235.1 | 4 | 1208 | ||
| GenBank mRNA | L09753.1 | - | 4 | 1030 | ||