siRNA Details
| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310.3 | NP_001301.1 | 4 | 2010 | ||
| GenBank mRNA | AF039081.1 | - | 4 | 1946 | ||
| AK002189.1 | - | 1 | 1027 | |||
| BC106052.1 | - | 4 | 1951 | 6150812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CREBL2| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310.3 | NP_001301.1 | 4 | 2010 | ||
| GenBank mRNA | AF039081.1 | - | 4 | 1946 | ||
| AK002189.1 | - | 1 | 1027 | |||
| BC106052.1 | - | 4 | 1951 | 6150812 | ||