siRNA Details
| Gene Symbol: DGKQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001347.3 | NP_001338.2 | 23 | 4397 | ||
| XM_011513411.1 | XP_011511713.1 | 23 | 4382 | |||
| GenBank mRNA | AB209503.1 | - | 23 | 4149 | ||
| BC063801.1 | - | 23 | 4406 | 6526640 | ||

DGKQ
Genomic Map