siRNA Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319165.1 | NP_001306094.1 | 4 | 1129 | ||
| NM_001319166.1 | NP_001306095.1 | 4 | 866 | |||
| NM_001319167.1 | NP_001306096.1 | 4 | 1196 | |||
| NM_001319168.1 | NP_001306097.1 | 4 | 1087 | |||
| NM_001319169.1 | NP_001306098.1 | 3 | 915 | |||
| NM_001319170.1 | NP_001306099.1 | 4 | 1199 | |||
| NM_001319171.1 | NP_001306100.1 | 4 | 1170 | |||
| NM_001384.4 | NP_001375.2 | 4 | 1139 | |||
| XM_017000502.1 | XP_016855991.1 | 4 | 1141 | |||
| XM_017000503.1 | XP_016855992.1 | 4 | 1075 | |||
| XM_017000504.1 | XP_016855993.1 | 3 | 1380 | |||
| XM_017000505.1 | XP_016855994.1 | 3 | 1312 | |||
| XM_017000506.1 | XP_016855995.1 | 4 | 1183 | |||
| GenBank mRNA | AF053003.1 | - | 4 | 1105 | ||
| AK091303.1 | - | 4 | 1058 | |||
| AK297933.1 | - | 3 | 888 | |||
| AK315506.1 | - | 4 | 1123 | |||
| BC001389.2 | - | 4 | 1098 | 3051455 | ||
| BC003181.1 | - | 4 | 1116 | 2958202 | ||
| BC016956.1 | - | 4 | 1098 | 3900854 | ||
| BT007431.1 | - | 4 | 944 | |||

DPH2
Genomic Map